missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Integra™ Miltex™ Halsted Mosquito Forceps

Serrations are full horizontal

$178.63 - $184.37

Specifications

Length (English) 3.87 in.
Length (Metric) 9.8cm
Product Type Hartman Mosquito Forceps
Quantity (No. per Case) Each
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Tip Style Price Quantity  
Catalog Number Mfr. No. Tip Style Price Quantity  
12-460-173 Integra™ Miltex™
MLT724
Straight Each for $178.63
12-460-174 Integra™ Miltex™
MLT726
Curved Each for $184.37
Specifications

Description

  • Pattern length 3.875 in. (9.8cm)
  • Available in curved and straight

Description & Specifications

Specifications

3.87 in.
9.8cm
Hartman Mosquito Forceps
Each