Please login to your online account to display your discounted pricing

Thermo Scientific™ Insulin Receptor (α -Subunit) Ab-3, Mouse Monoclonal Antibody

Recommended for a variety of molecular biology techniques, Thermo Scientific™ Insulin Receptor (α-Subunit) Ab-3, Mouse Monoclonal Antibody provides accurate, reproducible results.

$273.86 - $1,382.61

Specifications

Antigen Insulin Receptor (alpha-Subunit) Ab-3
Clone 47-9
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Bovine
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS633P

 
thermo scientific™
MS633P
200μg/mL 1mL Each for $1,282.55

MS633P0

 
thermo scientific™
MS633P0
200μg/mL 100µL Each for $273.86

MS633P1

 
thermo scientific™
MS633P1
200μg/mL 500µL Each for $797.16

MS633P1ABX

 
thermo scientific™
MS633P1ABX
1mg/mL 100µL Each for $877.06

MS633PABX

 
thermo scientific™
MS633PABX
1mg/mL 200µL Each for $1,382.61
Description & Specifications

Specifications

Antigen Insulin Receptor (alpha-Subunit) Ab-3
Clone 47-9
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Porcine
Cross Reactivity Sheep
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Porcine
Cross Reactivity Sheep
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Porcine
Cross Reactivity Sheep
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Porcine
Cross Reactivity Sheep
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline Steroids Hormones and Growth Factors
Type Primary
Immunogen IM-9 lymphocytes followed by purified insulin receptor
Isotype IgG1

Recommended for a variety of molecular biology techniques, Thermo Scientific™ Insulin Receptor (α-Subunit) Ab-3, Mouse Monoclonal Antibody provides accurate, reproducible results.

The human insulin receptor is a heterotetrameric membrane glycoprotein consisting of disulfide-linked subunits in a Β-α-α-Β configuration. The Β-subunit (95kDa) possesses a single transmembrane domain with tyrosine kinase activity, whereas the α-subunit (135kDa) is completely extracellular.

Host Species: Mouse

Clone: 47-9

Isotype: IgG1

Species Reactivity: Human, Cow, Pig and Sheep. Weakly reacts with Rabbit. Does not react with mouse and rat. Others not known.

Epitope: Exon 7/8

Immunogen: IM-9 lymphocytes followed by purified insulin receptor

Molecular Weight: 135kDa

Positive Control: IM-9 lymphocyte cells

Cellular Localization: Membrane Ab-3 inhibits (>90%) insulin binding and Blocks insulin action.

Recommended for:

  • Western Blotting (Not suitable)
  • Immunoprecipitation (Not suitable)
  • Antagonist (Blocks insulin action)
  • Completely Inhibits (>90%) Insulin Binding

Note:

USA: RUO; Int'l: RUO