Learn More
Insulin Receptor (α-Subunit) Ab-1 Mouse Monoclonal Antibody, Epredia™
Epredia™ Insulin Receptor (α-Subunit) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.
$369.67 - $1885.19
Specifications
Antigen | Insulin Receptor (alpha-Subunit) Ab-1 |
---|---|
Clone | 83-14 |
Applications | Immunoprecipitation |
Classification | Monoclonal |
Form | Concentrated |
Catalog Number | Mfr. No. | Conjugate | Concentration | Quantity | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Conjugate | Concentration | Quantity | Price | Quantity | |||
MS631P0
|
Thermo Fisher Scientific
MS631P0 |
Unconjugated | 200μg/mL | 100 μL |
Each for $369.67
|
|
|||
MS631P1
|
Thermo Fisher Scientific
MS631P1 |
Unconjugated | 200μg/mL | 500 μL |
Each for $1,086.75
|
|
|||
MS631P1ABX
|
Thermo Fisher Scientific
MS631P1ABX |
Unconjugated | 1mg/mL | 100 μL |
Each for $1,302.19
|
|
|||
MS631PABX
|
Thermo Fisher Scientific
MS631PABX |
Unconjugated | 1mg/mL | 200 μL |
Each for $1,885.19
|
|
|||
Description
The human insulin receptor is a heterotetrameric membrane glycoprotein consisting of disulfide-linked subunits in a Β-α-α-Β configuration. The Β-subunit (95kDa) possesses a single transmembrane domain, whereas the α-subunit (135kDa) is completely extracellular.Host Species: Mouse
Clone: 83-14
Isotype: IgG2a/κ
Species Reactivity: Human. Weakly with Cow, Pig and Sheep. Does not react with rabbit, mouse and rat. Others not known.
Epitope: aa 469-592 (exon 7/8)
Immunogen: IM-9 lymphocytes followed by purified insulin receptor
Molecular Weight: 135kDa
Positive Control: IM-9 lymphocyte cells, Placenta
Cellular Localization: Membrane Ab-1 inhibits insulin binding and mimics insulin action. It is excellent for immunoprecipitation of alpha-subunit of IR.
Recommended for:
- Immunoprecipitation
- ELISA
- Insulin-like Agonist
- Inhibits (∽80%) Insulin Binding (10nM)
- Tyrosine Kinase Assay (Ab-mediated capture on microplates)
Note:
USA: RUO; Int'l: RUO
Specifications
Insulin Receptor (alpha-Subunit) Ab-1 | |
Immunoprecipitation | |
Concentrated | |
RUO | |
Human | |
IgG2a κ |
83-14 | |
Monoclonal | |
Mouse | |
Steroids Hormones and Growth Factors | |
IM-9 lymphocytes followed by purified insulin receptor | |
Primary |