Please login to your online account to display your discounted pricing

Insulin Mouse, Biotin, Clone: ICBTACLS, eBioscience™

Mouse Monoclonal Antibody

$175.45 - $175.45

Specifications

Antigen Insulin
Clone ICBTACLS
Host Species Mouse
Species Reactivity Human, Mouse, Rat
Applications Immunohistochemistry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122608

 
life technologies
LS13976980
25μg Each for $175.45
Description & Specifications

Specifications

Antigen Insulin
Clone ICBTACLS
Host Species Mouse
Species Reactivity Human, Mouse, Rat
Applications Immunohistochemistry
Applications Immunohistochemistry (Formalin/Paraffin)
Applications Microscopy
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

This ICBTACLS monoclonal antibody reacts with the hormone insulin, responsible for regulation of carbohydrate and fat metabolism. Blood glucose levels are maintained by insulin. Insulin secretion from pancreatic beta-cells within the islets of Langerhans results in uptake of excess glucose from the blood into organs and its storage in the form of glycogen. Patients with type 1 diabetes have an autoimmune destruction of insulin-producing beta-cells in the pancreas, resulting in insulin deficiency and elevated levels of blood glucose. Patients with type 2 diabetes have insulin resistance where insulin becomes less effective at reducing the amount of blood glucose. Insulin is also important for proper endothelial cell function in with respect to vascular elasticity in response to changing blood pressure and plays a role in cognition, learning, and memory.

The ICBTACLS antibody recognizes human, mouse, and rat insulin.