Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Insulin Ab-5, Mouse Monoclonal Antibody

Ideal for Immunohistochemistry (Formalin/paraffin), Thermo Scientific™ Insulin Ab-5, Mouse Monoclonal Antibody provides accurate, reproducible results.

$240.37 - $755.90

Specifications

Antigen Insulin Ab-5
Clone INS05; same as 2D11-H5
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

MS1378P

 
thermo scientific™ lab vision™
MS1378P
Purified Ab with BSA and Azide 1mL Each for $660.65

MS1378P0

 
thermo scientific™ lab vision™
MS1378P0
Purified Ab with BSA and Azide 100μL Each for $240.37

MS1378P1

 
thermo scientific™ lab vision™
MS1378P1
Purified Ab with BSA and Azide 500μL Each for $474.39

MS1378P1ABX

 
thermo scientific™ lab vision™
MS1378P1ABX
Ab Without BSA/Azide (Frozen) 100μL Each for $603.10

MS1378PABX

 
thermo scientific™ lab vision™
MS1378PABX
Ab Without BSA/Azide (Frozen) 200μL Each for $755.90
Description & Specifications

Specifications

Antigen Insulin Ab-5
Clone INS05; same as 2D11-H5
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Porcine
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Porcine
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Porcine
Format Concentrated
Host Species Murine
Regulatory Status RUO
Research Discipline Steroids Hormones and Growth Factors
Type Primary
Immunogen Purified Insulin protein
Isotype IgG2a kappa

Ideal for Immunohistochemistry (Formalin/paraffin), Thermo Scientific™ Insulin Ab-5, Mouse Monoclonal Antibody provides accurate, reproducible results.

Insulin homeostasis; Insulin is a 51-amino acid polypeptide composed of A and B chains connected through the C-peptide. Proinsulin, which has very little biological activity, is cleaved by proteases within its cell of origin into the insulin molecule and the C-terminal basic residue. Insulin enhances membrane transport of glucose, amino acids, and certain ions. It also promotes glycogen storage, formation of triglycerides, and synthesis of proteins and nucleic acids. The main storage site for insulin is the pancreatic islets. Antibodies to insulin are important as β-cell and tumor (insulinoma) marker.

Host Species: Mouse

Clone: INS05; same as 2D11-H5

Isotype: IgG1/κ

Species Reactivity: Human, Pig and Rat. Others not known.

Epitope: Not determined

Immunogen: Purified Insulin protein

Molecular Weight: 6kDa

Positive Control: Pancreas

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO