Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Insulin Ab-5, Mouse Monoclonal Antibody


Ideal for Immunohistochemistry (Formalin/paraffin), Thermo Scientific™ Insulin Ab-5, Mouse Monoclonal Antibody provides accurate, reproducible results.

Manufacturer: thermo scientific™ lab vision™  MS1378P1ABX

 View more versions of this product

Catalog No. MS1378P1ABX

  • / Each
 

Description & Specifications

Specifications

Antigen Insulin Ab-5
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone INS05; same as 2D11-H5
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Porcine
Format Concentrated
Formulation Ab Without BSA/Azide (Frozen)
Host Species Murine
Immunogen Purified Insulin protein
Isotype IgG2a kappa
Quantity 100μL
Regulatory Status RUO
Research Discipline Steroids Hormones and Growth Factors
Type Primary
Monoclonal or Polyclonal Monoclonal
Target Species Canine
Target Species Human
Target Species Porcine

Ideal for Immunohistochemistry (Formalin/paraffin), Thermo Scientific™ Insulin Ab-5, Mouse Monoclonal Antibody provides accurate, reproducible results.

Insulin homeostasis; Insulin is a 51-amino acid polypeptide composed of A and B chains connected through the C-peptide. Proinsulin, which has very little biological activity, is cleaved by proteases within its cell of origin into the insulin molecule and the C-terminal basic residue. Insulin enhances membrane transport of glucose, amino acids, and certain ions. It also promotes glycogen storage, formation of triglycerides, and synthesis of proteins and nucleic acids. The main storage site for insulin is the pancreatic islets. Antibodies to insulin are important as β-cell and tumor (insulinoma) marker.

Host Species: Mouse

Clone: INS05; same as 2D11-H5

Isotype: IgG1/κ

Species Reactivity: Human, Pig and Rat. Others not known.

Epitope: Not determined

Immunogen: Purified Insulin protein

Molecular Weight: 6kDa

Positive Control: Pancreas

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO