Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Insulin Ab-5, Mouse Monoclonal Antibody


Ideal for Immunohistochemistry (Formalin/paraffin), Thermo Scientific™ Insulin Ab-5, Mouse Monoclonal Antibody provides accurate, reproducible results.

Manufacturer: thermo scientific™ lab vision™  MS1378PABX

 View more versions of this product

Catalog No. MS1378PABX

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Porcine
Quantity 200µL
Isotype IgG2a kappa
Clone INS05; same as 2D11-H5
Immunogen Purified Insulin protein
Antigen Insulin Ab-5
Format Concentrated
Applications Immunohistochemistry (Paraffin)
Regulatory Status RUO
Conjugate Unlabeled
Type Primary
Classification Monoclonal
Formulation Ab Without BSA/Azide (Frozen)
Research Discipline Steroids Hormones and Growth Factors
Concentration 1mg/mL

Ideal for Immunohistochemistry (Formalin/paraffin), Thermo Scientific™ Insulin Ab-5, Mouse Monoclonal Antibody provides accurate, reproducible results.

Insulin homeostasis; Insulin is a 51-amino acid polypeptide composed of A and B chains connected through the C-peptide. Proinsulin, which has very little biological activity, is cleaved by proteases within its cell of origin into the insulin molecule and the C-terminal basic residue. Insulin enhances membrane transport of glucose, amino acids, and certain ions. It also promotes glycogen storage, formation of triglycerides, and synthesis of proteins and nucleic acids. The main storage site for insulin is the pancreatic islets. Antibodies to insulin are important as β-cell and tumor (insulinoma) marker.

Host Species: Mouse

Clone: INS05; same as 2D11-H5

Isotype: IgG1/κ

Species Reactivity: Human, Pig and Rat. Others not known.

Epitope: Not determined

Immunogen: Purified Insulin protein

Molecular Weight: 6kDa

Positive Control: Pancreas

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO