Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Insulin Ab-4, Mouse Monoclonal Antibody

Ideal for Immunohistochemistry (Formalin/paraffin), Thermo Scientific™ Insulin Ab-4, Mouse Monoclonal Antibody provides accurate, reproducible results.

$235.69 - $984.35

Specifications

Antigen Insulin Ab-4
Clone INS04; same as E2-E3
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Cross Reactivity Human
View More Specs
Products
Catalog Number Mfr. No. Concentration Conjugate Quantity Price Quantity    

MS1377B

 
thermo scientific™ lab vision™
MS1377B
200μg/mL Biotin 1mL Each for $984.35

MS1377B0

 
thermo scientific™ lab vision™
MS1377B0
200μg/mL Biotin 100µL Each for $331.51

MS1377B1

 
thermo scientific™ lab vision™
MS1377B1
200μg/mL Biotin 500µL Each for $707.76

MS1377P

 
thermo scientific™ lab vision™
MS1377P
200μg/mL Unlabeled 1mL Each for $654.06

MS1377P0

 
thermo scientific™ lab vision™
MS1377P0
200μg/mL Unlabeled 100µL Each for $235.69

MS1377P1

 
thermo scientific™ lab vision™
MS1377P1
200μg/mL Unlabeled 500µL Each for $470.14

MS1377P1ABX

 
thermo scientific™ lab vision™
MS1377P1ABX
1mg/mL Unlabeled 100µL Each for $590.41

MS1377PABX

 
thermo scientific™ lab vision™
MS1377PABX
1mg/mL Unlabeled 200µL Each for $734.33
Description & Specifications

Specifications

Antigen Insulin Ab-4
Clone INS04; same as E2-E3
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Cross Reactivity Human
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline Steroids Hormones and Growth Factors
Type Primary
Immunogen Swine insulin
Isotype IgG2a kappa

Ideal for Immunohistochemistry (Formalin/paraffin), Thermo Scientific™ Insulin Ab-4, Mouse Monoclonal Antibody provides accurate, reproducible results.

Insulin is a 51-amino acid polypeptide composed of A and B chains connected through the C-peptide. Proinsulin, which has very little biological activity, is cleaved by proteases within its cell of origin into the insulin molecule and the C-terminal basic residue. Insulin enhances membrane transport of glucose, amino acids, and certain ions. It also promotes glycogen storage, formation of triglycerides, and synthesis of proteins and nucleic acids. The main storage site for insulin is the pancreatic islets. Antibodies to insulin are important as β-cell of islets of Langerhans and tumor (insulinoma) marker.

Host Species: Mouse

Clone: INS04; same as E2-E3

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Swine insulin

Molecular Weight: 6kDa

Positive Control: Pancreas

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO