Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Insulin Ab-4, Mouse Monoclonal Antibody


Ideal for Immunohistochemistry (Formalin/paraffin), Thermo Scientific™ Insulin Ab-4, Mouse Monoclonal Antibody provides accurate, reproducible results.

Manufacturer: thermo scientific™ lab vision™  MS1377P1ABX

 View more versions of this product

Catalog No. MS1377P1ABX

  • / Each
 

Description & Specifications

Specifications

Antigen Insulin Ab-4
Format Concentrated
Isotype IgG2a kappa
Quantity 100µL
Regulatory Status RUO
Cross Reactivity Human
Host Species Mouse
Applications Immunohistochemistry (Paraffin)
Immunogen Swine insulin
Clone INS04; same as E2-E3
Type Primary
Research Discipline Steroids Hormones and Growth Factors
Classification Monoclonal
Concentration 1mg/mL
Conjugate Unlabeled

Ideal for Immunohistochemistry (Formalin/paraffin), Thermo Scientific™ Insulin Ab-4, Mouse Monoclonal Antibody provides accurate, reproducible results.

Insulin is a 51-amino acid polypeptide composed of A and B chains connected through the C-peptide. Proinsulin, which has very little biological activity, is cleaved by proteases within its cell of origin into the insulin molecule and the C-terminal basic residue. Insulin enhances membrane transport of glucose, amino acids, and certain ions. It also promotes glycogen storage, formation of triglycerides, and synthesis of proteins and nucleic acids. The main storage site for insulin is the pancreatic islets. Antibodies to insulin are important as β-cell of islets of Langerhans and tumor (insulinoma) marker.

Host Species: Mouse

Clone: INS04; same as E2-E3

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Swine insulin

Molecular Weight: 6kDa

Positive Control: Pancreas

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO