Please login to your online account to display your discounted pricing
SHOWA™ Insulated Super Flex™ Gloves
Provide superior chemical and abrasion resistance
$11.01 - $11.01
Products
Catalog Number Mfr. No. Price Quantity    
29-635-7320
showa™
7510*
Pair for $11.01
Case of 48 Pair for $505.44
Description & Specifications
Gloves are recyclable and ideal for jobs in cold storage, outdoor work in cold climates and fishing.
  • PVC plastic coating and wrinkle finish allow excellent wet or dry grip
  • Foam insulation sandwiched between 100% cotton jersey and fully coated with triple-layered insulation protects from cold while locking in warmth
  • Gloves remain flexible even in subzero temperatures
  • Color: orange