Instant Ocean™ Synthetic Sea Salts
An easy sea salt alternative!
Manufacturer:  Fisher Science Education SS1200
Catalog No. S17957
Description
Description
  • Add to ordinary tap water to provide a culture medium that's considered superior to natural seawater
  • These salts do not imitate natural seawater, but rather produce a medium in which sea life thrives
Specifications
Specifications
50 gal.
185L
Produce a medium in which sea life thrives
Instant ocean synthetic sea Salt
Documents