LaMotte™ Insta-Test™ Water Test Strips
Perform safe, quick and easy water quality checks with test strips.
Manufacturer:  LaMotte™ 2994
 View more versions of this product
Catalog No. S66232
Description
Description
  • Monitor water without having to use reagents or field kits
Order Info
Each vial of test strips contains 50 tests per factor, except S66233, which contains 25 tests per factor.
Specifications
Specifications
Drinking, pool water testing
0 to 5ppm (Iron), 0 to 3ppm (Copper)
Education
25 Tests
Water Test Strips
Documents