LaMotte™ Insta-Test™ Water Test Strips
Perform safe, quick and easy water quality checks with Insta-Test™ Water Test Strips.
Supplier: LaMotte™ EPD2987G
Description
- Monitor water without having to use reagents or field kits.
Order Info
Each vial of test strips contains 50 tests per factor, except S66233, which contains 25 tests per factor.
Specifications
Total Chlorine, pH | |
Drinking, food/beverage, pool water testing | |
4 to 10 (pH), 0 to 50 (TCl) |
Strip | |
50 Pk. | |
Water Test Strip |