Invitrogen™ INSRR (IRR) Recombinant Human Protein DFS Item
INSRR, insulin receptor-related receptor, is a member of the insulin receptor (IR) family.
Manufacturer:  Invitrogen™ LSPV4111
 View more versions of this product
Catalog No. PV4111
Description
Description
INSRR does not bind insulin-like molecules, but is similar to INSR in tyrosine kinase activity.
Specifications
Specifications
NP_055030
Frozen
67.1kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Cytoplasmic
Kinase Assay
Insulin & IGF Receptor Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
INSRR (IRR), IRR
Baculovirus
Human
GST Tag
Documents