Invitrogen™ INSR Recombinant Human Protein DFS Item
INSR is a receptor tyrosine kinase.
Manufacturer:  Invitrogen™ LSPV4110
 View more versions of this product
Catalog No. PV4110
Description
Description
Binding of insulin to the extracellular domain of INSR stimulates glucose uptake.
Specifications
Specifications
NP_000199
Frozen
71.5kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Cytoplasmic
Kinase Assay
Insulin & IGF Receptor Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
INSR
Baculovirus
Human
GST Tag
Documents