missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Inosine, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 122251000
Specifications
1.65 to 1.77 (A250/A260), 0.22 to 0.26 (A280/A260) | |
Authentic | |
Glass bottle | |
-49.2 | |
0.1% max. | |
White | |
100g | |
98.5% min. (HPLC) | |
MFCD00066770 | |
15, 5018 | |
Solubility in water: 2.1g/100 mL (20°C). Other solubilities: soluble in ethanol | |
C1=NC(=O)C2=C(N1)N(C=N2)C3C(C(C(O3)CO)O)O | |
268.22 | |
CHEBI:17596 | |
Crystalline Powder |
Inosine | |
1% max. | |
− 49.20 (18.00°C c=1,H2O) | |
98 to 102% (lambda max. 248.5nm,pH 7) Molar absorptivity 12200+/-2%,on dry substance | |
98% min. (c=1, H2O, 430nm, 1cm cell) | |
212°C to 213°C | |
58-63-9 | |
C10H12N4O5 | |
31, 25 | |
atorel, hypoxanthine riboside, hypoxanthosine, inosine, oxiamin, panholic-l, pantholic-l, ribonosine, selfer, trophicardyl | |
UGQMRVRMYYASKQ-KQYNXXCUSA-N | |
9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one | |
6021 | |
268.22 | |
99% |
Chemical Identifiers
58-63-9 | |
268.22 | |
UGQMRVRMYYASKQ-KQYNXXCUSA-N | |
6021 | |
9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one |
C10H12N4O5 | |
MFCD00066770 | |
atorel, hypoxanthine riboside, hypoxanthosine, inosine, oxiamin, panholic-l, pantholic-l, ribonosine, selfer, trophicardyl | |
CHEBI:17596 | |
C1=NC(=O)C2=C(N1)N(C=N2)C3C(C(C(O3)CO)O)O |
Safety and Handling
EINECSNumber : 200-390-4