Please login to your online account to display your discounted pricing

Alfa Aesar™ Inosine-5'-monophosphoric acid disodium salt hydrate, 99%

$151.96 - $151.96

Chemical Identifiers

Chemical Name or Material Inosine-5'-monophosphoric acid disodium salt hydrate
CAS 352195-40-5
Molecular Formula C10H11N4Na2O8P·xH2O
Formula Weight 392.24g/mol (Anhydrous)
MDL Number MFCD00036201
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1072914

 
alfa aesar™
A10729-14
25g Each for $151.96
Description & Specifications

Specifications

Quantity 25g
Chemical Name or Material Inosine-5'-monophosphoric acid disodium salt hydrate
CAS 352195-40-5
Assay 99%
Molecular Formula C10H11N4Na2O8P·xH2O
Formula Weight 392.24g/mol (Anhydrous)
MDL Number MFCD00036201
Recommended Storage 2°C to 8°C
RTECS Number NM7519500
TSCA TSCA
Safety and Handling

2°C to 8°C