missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Inosine 5′-Monophosphate Disodium Salt Hydrate, TCI America™

$21.39 - $125.28

Chemical Identifiers

CAS 4691-65-0
Molecular Formula C10H13N4Na2O8P
Molecular Weight (g/mol) 394.187
MDL Number MFCD00036201
InChI Key SLIUELDPAPMQIB-IDIVVRGQSA-N
Synonym disodium 5'-inosinate
PubChem CID 123134061
IUPAC Name [(2R,3S,4R,5R)-3,4-dihydroxy-5-(6-oxo-3H-purin-9-yl)oxolan-2-yl]methyl dihydrogen phosphate;sodium
SMILES C1=NC(=O)C2=C(N1)N(C=N2)C3C(C(C(O3)COP(=O)(O)O)O)O.[Na].[Na]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I003625G View Documents TCI America
I003625G
25g Each for $125.28
 
I00361G View Documents TCI America
I00361G
1g Each for $21.39
 
I00365G View Documents TCI America
I00365G
5g Each for $42.78
 
Specifications

Chemical Identifiers

4691-65-0
394.187
SLIUELDPAPMQIB-IDIVVRGQSA-N
123134061
C1=NC(=O)C2=C(N1)N(C=N2)C3C(C(C(O3)COP(=O)(O)O)O)O.[Na].[Na]
C10H13N4Na2O8P
MFCD00036201
disodium 5'-inosinate
[(2R,3S,4R,5R)-3,4-dihydroxy-5-(6-oxo-3H-purin-9-yl)oxolan-2-yl]methyl dihydrogen phosphate;sodium

Specifications

White
4691-65-0
MFCD00036201
SLIUELDPAPMQIB-IDIVVRGQSA-N
[(2R,3S,4R,5R)-3,4-dihydroxy-5-(6-oxo-3H-purin-9-yl)oxolan-2-yl]methyl dihydrogen phosphate;sodium
123134061
Crystal-Powder at 20°C
Inosine 5′-Monophosphate Disodium Salt Hydrate
25g
C10H13N4Na2O8P
disodium 5'-inosinate
C1=NC(=O)C2=C(N1)N(C=N2)C3C(C(C(O3)COP(=O)(O)O)O)O.[Na].[Na]
394.187
392.17
≥98.0% (HPLC,E)
Safety and Handling

Safety and Handling

EINECSNumber : (8)-0276

RTECSNumber : NM7519500

TSCA : Yes