missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Innovating Science™ Blood Typing Anti-Sera
For blood typing using real human blood. 5 mL vials contain enough antisera for 75 students. Sample will keep over one year with proper refrigeration.
Manufacturer: Innovating Science™ IS3151
Description
- Five milliliter vial contains enough antisera for 75 students
- Sample will keep over one year with proper refrigeration
- Materials from human-derived cell lines were found to be negative in tests for Hepatitis B, HIV and EBV
- Includes Anti-A & Anti-B, Anti Rh and Anti-H Lectin Anti-sera
DOT: Non regulated
Specifications
Ambient | |
1 | |
Anti-A and Anti-B and Anti Rh Anti-sera | |
Blood Typing Anti-Sera Type A, B and Rh | |
3 x .0.5 x 0.5 in. |
Educational | |
For Blood typing of real Human Blood | |
Plastic Bottle | |
1 |
For educational use only. Not FDA approved for clinical use.