Please login to your online account to display your discounted pricing
Novus Biologicals™ InnovaGOLD Gold Nanoparticles Antibody Labeling Kit, 40nm, 10 OD
$298.20 - $298.20
Products
Catalog Number Mfr. No. Quantity Price Quantity    
2100010
novus biologicals™
2100010
10mL Each for $298.20
Description & Specifications
  • Uniform Shape
  • Low co-efficient of variation in terms of size
  • High efficiency antibody binding for maximum sensitivity
  • Available at high concentrations to simplify assay development