Please login to your online account to display your discounted pricing
Novus Biologicals™ InnovaCoat™ GOLD Streptavidin Antibody Labeling Kit, 40nm
Gold nanoparticle with a unique protective surface coat. Covalently attached streptavidin ligand cannot desorb.
$870.00 - $870.00
Catalog Number Mfr. No. Quantity Price Quantity    
novus biologicals™
1mL Each for $870.00
Description & Specifications
  • Can withstand the most extreme conditions (2.5M NaH at 70°C for ≈2 hours)
  • Streptavidin is attached to InnovaCoat™ GOLD via an extended linker to facilitate high-affinity molecular interactions with avidin and streptavidin