Please login to your online account to display your discounted pricing
Novus Biologicals™ InnovaCoat™ GOLD Hydrazide Antibody Labeling Kit, 40nm
Quick and easy attachment of antibodies. Hydrazide groups bind antibodies via the Fc region, orienting the two antigen-binding arms in space.
$747.59 - $747.59
Products
Catalog Number Mfr. No. Quantity Price Quantity    
2800100
novus biologicals™
2800100
1 x 10-30μg Reaction Each for $747.59
Description & Specifications
Features
  • Proprietary surface coat
  • Ultra-stable
  • Covalently link antibodies and analytes
  • Simple conjugation processes
  • Different surface chemistries
Benefits
  • Enhanced assay sensitivity
  • Protective coat prevents metal-antibody interactions
  • Rapid screening of multiple antibodies for assay development
  • Reduced antibody loading without aggregation
  • No pH titrations
  • Fully scalable – from R&D to production
  • Custom InnovaCoat™ labeling services