Please login to your online account to display your discounted pricing
Novus Biologicals™ InnovaCoat™ GOLD Biotin Antibody Labeling Kit, 40nm
Gold nanoparticle with a unique protective surface coat. Covalently attached biotin ligand cannot desorb.
$846.29 - $846.29
Products
Catalog Number Mfr. No. Quantity Price Quantity    
2401000
novus biologicals™
2401000
1mL Each for $846.29
Description & Specifications
  • Can withstand the most extreme conditions (2.5M NaH at 70°C for ≈2 hours)
  • Biotin is attached to InnovaCoat™ GOLD via an extended linker to facilitate high-affinity molecular interactions with avidin and streptavidin