missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Innova™ 42R Shaker and Universal Platform Bundle PROMO
The Innova™ 42R refrigerated shaker plus 18 x 18 in. universal platform speeds plasmid prep, cell culture or enzyme screens. Stack units to free space and run multi-vessel workflows with ease.
$30156.56 - $30156.56
Specifications
| Load Bearing Capacity (Metric) | 9 to 15.8 kg |
|---|---|
| Speed Range | 25 to 400 rpm |
| Electrical Requirements | 120 V 50/60 Hz |
| Voltage | 120 V |
| Temperature Range (Metric) | 20°C to 80°C |
| Catalog Number | Mfr. No. | Orbit | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Orbit | Price | Quantity | |||||
|
05400157PM1
|
Eppendorf™
2231001156 |
19 mm |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
05400162PM1
|
Eppendorf™
2231001157 |
25 mm |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Innova™ 42R shaker and Universal Platform Bundle Includes:
- 1 x Innova™ 42R with 3/4 in. or 1 in. orbit, 120V/60Hz
- 1 x Universal Platforms, 18 x 18 in.
Innova™ 42R Shaker Features:
- Height adjustable static shelf allows simultaneous shaking and static culture with different vessels
- Save space by stacking units or positioning them on or under a standard-height lab bench
- Benefit from configurable platforms that accommodate tubes, microplates, and flasks up to 6 L
Specifications
| 9 to 15.8 kg | |
| 120 V 50/60 Hz | |
| 20°C to 80°C | |
| 6 L Flasks | |
| 20 to 35 lb. | |
| ±0.25°C at 37°C | |
| Aluminum | |
| 20.3 in. | |
| 51.5 cm | |
| 46 x 46 cm (18 x 18 in.) | |
| 32.2 in. | |
| 81.8 cm | |
| 25 in. | |
| 63.5 cm | |
| 266 lb. | |
| ±1 rpm | |
| LED Digital Electric Display | |
| Universal/Dedicated | |
| Yes |
| 25 to 400 rpm | |
| 120 V | |
| 12.5 A | |
| Incubator Shaker | |
| 68°F to 176°F | |
| 60 Hz | |
| 29.3 in. | |
| 74.5 cm | |
| 51.5 x 51.7 x 47.7 cm (20.3 x 20.4 x 18.8 in.) | |
| 63.5 x 74.5 x 81.8 cm (25 x 29.3 x 32.8 in.) | |
| 18.8 in. | |
| 47.7 cm | |
| 20.4 in. | |
| 51.7 cm | |
| 121 kg | |
| Innova 42R | |
| 20 to 80%, Non-condensing | |
| 0.01 sec. to 95 hr. 59 min., Continuous |