Please login to your online account to display your discounted pricing

INH1, Tocris™

Hec1 inhibitor; causes arrest of mitosis

$208.25 - $867.95

Chemical Identifiers

Chemical Name or Material INH1
Assay Percent Range >99%
Molecular Formula C18H16N2OS·HCl
Formula Weight 344.86
Synonym N-[4-(2, 4-Dimethylphenyl)-2-thiazolyl]benzamide hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

355110

 
r & d systems
RDS355110
10mg Each for $208.25

355150

 
r & d systems
RDS355150
50mg Each for $867.95
Description & Specifications

Specifications

Formula Weight 344.86
Quantity 10mg
Chemical Name or Material INH1
Assay Percent Range >99%
Molecular Formula C18H16N2OS·HCl
CAS 313553-47-8
Synonym N-[4-(2, 4-Dimethylphenyl)-2-thiazolyl]benzamide hydrochloride
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C