missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Infusorial Earth (Pinkish to White Hue Powder ), Fisher Chemical

Manufacturer:  Fisher Chemical I223

Catalog No. I22-3



Description

Description

Specifications

Specifications

Infusorial Earth
O2Si
aerosil, cristobalite, diatomaceous earth, infusorial earth, quartz, silica, silica, amorphous, silicic anhydride, silicon dioxide, tridymite
O=[Si]=O
60.083
CHEBI:30563
2.3g/cm³
Poly Pail
9 to 10
61790-53-2
MFCD00132803
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
dioxosilane
24261
Solid
Pass test
White
3kg

Chemical Identifiers

61790-53-2
60.083
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
24261
dioxosilane
O2Si
MFCD00132803
aerosil, cristobalite, diatomaceous earth, infusorial earth, quartz, silica, silica, amorphous, silicic anhydride, silicon dioxide, tridymite
CHEBI:30563
O=[Si]=O
Safety and Handling

Safety and Handling

SDS Documents