Please login to your online account to display your discounted pricing

Alfa Aesar™ Influenza A NP (366-374)Strain A/PR/8/35

For cellular and molecular biology applications

Specifications

Product Type Influenza A NP (366-374) Strain A/PR/8/35
Molecular Weight 1026.10
Storage Requirements -30° to -10°C
Format Solid
Sequence Ala-Ser-Asn-Glu-Asn-Met-Glu-Thr-Met
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66783MA

 
alfa aesar™
J66783MA
10mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66783MB

 
alfa aesar™
J66783MB
25mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Influenza A NP (366-374) Strain A/PR/8/35
Molecular Weight 1026.10
Storage Requirements -30° to -10°C
Format Solid
Sequence Ala-Ser-Asn-Glu-Asn-Met-Glu-Thr-Met
Regulatory Status RUO
Purification Method HPLC