Please login to your online account to display your discounted pricing

Alfa Aesar™ Influenza A NP (366-374)

For cellular and molecular biology applications

Specifications

Product Type Influenza A NP (366-374)
Molecular Weight 982.05
Storage Requirements -30° to -10°C
Sequence Ala-Ser-Asn-Glu-Met-Asn-Asp-Ala-Met
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66443MA

 
alfa aesar™
J66443MA
10mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66443MB

 
alfa aesar™
J66443MB
25mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Influenza A NP (366-374)
Molecular Weight 982.05
Storage Requirements -30° to -10°C
Sequence Ala-Ser-Asn-Glu-Met-Asn-Asp-Ala-Met
Format Solid
Regulatory Status RUO
Purification Method HPLC