Please login to your online account to display your discounted pricing

Thermo Scientific™ Infinity™ ACE Assay


Measure angtiotensin-converting enzyme in serum with the Thermo Scientific™ Infinity™ ACE Assay.

Manufacturer: thermo scientific™  TR85201

 View more versions of this product

Catalog No. TR85201

  • / Each
 

Description & Specifications

Specifications

Detectable Analytes ACE (Angiotensin Converting Enzyme)
Detectable Analytes For use with the ACE reagent kit
Type Calibrator
Sample Type Serum
Methods Compatibility Kinetic
CE Marker Yes
Linearity 1 to 120U/L
Stability Stable Liquid
Description ACE Calibrator
Storage Requirements 2° to 8°C

Measure angtiotensin-converting enzyme in serum with Thermo Scientific™ Infinity™ ACE Assay, a liquid stable reagent used to aid in the differential diagnosis of clinically active pulmonary sarcoidosis and for monitoring the effectiveness of steroid therapy.

  • Used for monitoring the effects of ACE inhibitors in the treatment of hypertension and heart failure
  • Applications available for most automated chemistry analyzers