missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Infinity™ ACE Assay

Measure angtiotensin-converting enzyme in serum with the Thermo Scientific™ Infinity™ ACE Assay.

$336.71 - $642.03

Specifications

CE Marker Yes
Linearity 1 to 120U/L
Methods Compatibility Kinetic
Stability Stable Liquid
Storage Requirements 2° to 8°C
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
TR85056 Thermo Scientific™
TR85056
ACE (Angiotensin Converting Enzyme) Assay Each for $642.03
TR85101 Thermo Scientific™
TR85101
ACE Control Set Each for $336.71
TR85201 Thermo Scientific™
TR85201
ACE Calibrator Each for $347.10
Specifications

Description

Measure angtiotensin-converting enzyme in serum with Thermo Scientific™ Infinity™ ACE Assay, a liquid stable reagent used to aid in the differential diagnosis of clinically active pulmonary sarcoidosis and for monitoring the effectiveness of steroid therapy.

  • Used for monitoring the effects of ACE inhibitors in the treatment of hypertension and heart failure
  • Applications available for most automated chemistry analyzers
Description & Specifications

Specifications

Yes
Kinetic
2° to 8°C
1 to 120U/L
Stable Liquid