Please login to your online account to display your discounted pricing

Thermo Scientific™ Infinity™ ACE Assay


Measure angtiotensin-converting enzyme in serum with the Thermo Scientific™ Infinity™ ACE Assay.

Manufacturer: thermo scientific™  TR85056

 View more versions of this product

Catalog No. TR85056

  • / Each
 

Description & Specifications

Specifications

Detectable Analytes ACE (Angiotensin Converting Enzyme)
Detectable Analytes Used to aid in the differential diagnosis of clinically active pulmonary sarcoidosis and for monitoring the effectiveness of steroid therapy
Sample Type Serum
Type Reagent
Storage Requirements 2° to 8°C
Kit Size 2 x 28mL
Linearity 1 to 120U/L
CE Marker Yes
Description ACE (Angiotensin Converting Enzyme) Assay
Stability Stable Liquid
State at Room Temperature Liquid
Methods Compatibility Kinetic

Measure angtiotensin-converting enzyme in serum with Thermo Scientific™ Infinity™ ACE Assay, a liquid stable reagent used to aid in the differential diagnosis of clinically active pulmonary sarcoidosis and for monitoring the effectiveness of steroid therapy.

  • Used for monitoring the effects of ACE inhibitors in the treatment of hypertension and heart failure
  • Applications available for most automated chemistry analyzers