missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Indoxyl acetate, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC156660250

 View more versions of this product

Catalog No. AC156660250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Indoxyl acetate
Glass bottle
128.0°C to 131.0°C
608-08-2
C10H9NO2
21, 71
Solubility in water: insoluble. Other solubilities: soluble in ethanol,chloroform
CC(=O)OC1=CNC2=CC=CC=C21
175.19
175.19
97%
Authentic
Colorless to Lilac
25g
96% min. (GC)
MFCD00014561
1h-indol-3-ol, 3-acetate
JBOPQACSHPPKEP-UHFFFAOYSA-N
1H-indol-3-yl acetate
11841
Glistening Crystals

Chemical Identifiers

608-08-2
175.19
JBOPQACSHPPKEP-UHFFFAOYSA-N
11841
CC(=O)OC1=CNC2=CC=CC=C21
C10H9NO2
MFCD00014561
1h-indol-3-ol, 3-acetate
1H-indol-3-yl acetate
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS Signal Word: Warning

EINECSNumber : 210-154-2

SDS Documents