missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Indoline-2-carboxylic acid, 97%, Maybridge™

Chemical Identifiers

CAS 78348-24-0
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.176
MDL Number MFCD00010635
InChI Key QNRXNRGSOJZINA-UHFFFAOYSA-N
Synonym +/--indoline-2-carboxylic acid, 1-indoline-2-carboxylic acid, 1h-indole-2-carboxylic acid, 2,3-dihydro, 1h-indole-2-carboxylicacid, 2,3-dihydro, 2,3-dihydroindole-2-carboxylic acid, 2-indolinecarboxylic acid, dl-indoline-2-carboxylic acid, indoline-2-carboxylic acid, indoline-2-carboxylicacid, pubchem3035
PubChem CID 86074
IUPAC Name 2,3-dihydro-1H-indole-2-carboxylic acid
SMILES C1C(NC2=CC=CC=C21)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
BTB14322DA
SDS View Documents
Fisher Scientific Company
BTB14322DA
1g Amber Glass Bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BTB14322EA
SDS View Documents
Fisher Scientific Company
BTB14322EA
10g Amber Glass Bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

78348-24-0
163.176
QNRXNRGSOJZINA-UHFFFAOYSA-N
86074
C1C(NC2=CC=CC=C21)C(=O)O
C9H9NO2
MFCD00010635
+/--indoline-2-carboxylic acid, 1-indoline-2-carboxylic acid, 1h-indole-2-carboxylic acid, 2,3-dihydro, 1h-indole-2-carboxylicacid, 2,3-dihydro, 2,3-dihydroindole-2-carboxylic acid, 2-indolinecarboxylic acid, dl-indoline-2-carboxylic acid, indoline-2-carboxylic acid, indoline-2-carboxylicacid, pubchem3035
2,3-dihydro-1H-indole-2-carboxylic acid

Specifications

Amber Glass Bottle
78348-24-0
100.0
C9H9NO2
+/--indoline-2-carboxylic acid, 1-indoline-2-carboxylic acid, 1h-indole-2-carboxylic acid, 2,3-dihydro, 1h-indole-2-carboxylicacid, 2,3-dihydro, 2,3-dihydroindole-2-carboxylic acid, 2-indolinecarboxylic acid, dl-indoline-2-carboxylic acid, indoline-2-carboxylic acid, indoline-2-carboxylicacid, pubchem3035
C1C(NC2=CC=CC=C21)C(=O)O
163.176
163.18
Indoline-2-carboxylic acid
1g
96.5
97%
MFCD00010635
QNRXNRGSOJZINA-UHFFFAOYSA-N
2,3-dihydro-1H-indole-2-carboxylic acid
86074
97%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.