Please login to your online account to display your discounted pricing

Alfa Aesar™ Indolicidin

For cellular and molecular biology applications

Specifications

Product Type Indolicidin
Molecular Weight 1906.28
Storage Requirements -30° to -10°C
Sequence Ile-Leu-Pro-Trp-Lys-Trp-Pro-Trp-Trp-Pro-Trp-Arg-Arg-NH2
Format Powder
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ65215EXD

 
alfa aesar™
J65215EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ65215MCR

 
alfa aesar™
J65215MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Indolicidin
Molecular Weight 1906.28
Storage Requirements -30° to -10°C
Sequence Ile-Leu-Pro-Trp-Lys-Trp-Pro-Trp-Trp-Pro-Trp-Arg-Arg-NH2
Format Powder
Regulatory Status RUO
Purification Method HPLC