missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Indole, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 122150250

 View more versions of this product

Catalog No. AC122150250



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Indole
120-72-9
Pink to White
253.0°C to 254.0°C
Authentic
Glass bottle
MFCD00005607
20, 304
15, 5003
Solubility in water: 2.80g/L (25°C). Other solubilities: soluble in hot water,soluble in hot alcohol,ether and benzene,0.1g/mL meoh
C1=CC=C2C(=C1)C=CN2
117.15
CHEBI:16881
99+%
98+%
51.0°C to 53.0°C
1.2200g/mL
136°C
99+%
C8H7N
25 g
1.22
indole, indol, 2,3-benzopyrrole, 1-benzazole, ketole, 1-azaindene, benzopyrrole, 2,3-benzopyrole, caswell no. 498b, indol german
SIKJAQJRHWYJAI-UHFFFAOYSA-N
1H-indole
798
117.15
Crystalline Powder

Chemical Identifiers

120-72-9
SIKJAQJRHWYJAI-UHFFFAOYSA-N
798
1H-indole
MFCD00005607
indole, indol, 2,3-benzopyrrole, 1-benzazole, ketole, 1-azaindene, benzopyrrole, 2,3-benzopyrole, caswell no. 498b, indol german
CHEBI:16881
C1=CC=C2C(=C1)C=CN2
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Causes serious eye damage.
May cause an allergic skin reaction.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water

GHS Signal Word: Danger

EINECSNumber : 204-420-7

RTECSNumber : NL2450000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only