missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Indole, 99+%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 122150100
Specifications
Indole | |
136°C | |
Glass Bottle | |
253.0°C to 254.0°C | |
51.0°C to 53.0°C | |
98+% | |
99+% | |
MFCD00005607 | |
15, 5003 | |
Solubility in water: 2.80g/L (25°C). Other solubilities: soluble in hot water,soluble in hot alcohol,ether and benzene,0.1g/mL meoh | |
C1=CC=C2C(=C1)C=CN2 | |
117.15 | |
CHEBI:16881 | |
Crystalline Powder |
1.2200g/mL | |
Authentic | |
1.22 | |
Pink to White | |
10g | |
120-72-9 | |
C8H7N | |
20, 304 | |
1-azaindene, 1-benzazole, 2,3-benzopyrole, 2,3-benzopyrrole, benzopyrrole, caswell no. 498b, indol, indol german, indole, ketole | |
SIKJAQJRHWYJAI-UHFFFAOYSA-N | |
1H-indole | |
798 | |
117.15 | |
99+% |
Chemical Identifiers
120-72-9 | |
117.15 | |
SIKJAQJRHWYJAI-UHFFFAOYSA-N | |
798 | |
1H-indole |
C8H7N | |
MFCD00005607 | |
1-azaindene, 1-benzazole, 2,3-benzopyrole, 2,3-benzopyrrole, benzopyrrole, caswell no. 498b, indol, indol german, indole, ketole | |
CHEBI:16881 | |
C1=CC=C2C(=C1)C=CN2 |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Causes serious eye damage.
May cause an allergic skin reaction.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water
GHS Signal Word: Danger
EINECSNumber : 204-420-7
RTECSNumber : NL2450000
TSCA : TSCA