missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Indole-7-carboxylic acid, 97%

Manufacturer:  Alfa Aesar™ H5493114

 View more versions of this product

Catalog No. AAH5493114



Description

Description

Applications
Indole-7-carboxylic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

Indole-7-carboxylic acid
1670-83-3
MFCD00210442
Soluble in water.
C1=CC2=C(C(=C1)C(=O)O)NC=C2
161.16
161.05
25g
C9H7NO2
1h-indole-7-carboxylicacid, 7-carboxyindole, 7-indolecarboxylic acid, acmc-209dvl, indole 7-carboxylic acid, indole-7-carboxlic acid, indole-7-carboxylic acid, indole-7-carboxylicacid, pubchem2475, zlchem 444
IPDOBVFESNNYEE-UHFFFAOYSA-N
1H-indole-7-carboxylic acid
74281
97%

Chemical Identifiers

1670-83-3
161.16
IPDOBVFESNNYEE-UHFFFAOYSA-N
74281
C1=CC2=C(C(=C1)C(=O)O)NC=C2
C9H7NO2
MFCD00210442
1h-indole-7-carboxylicacid, 7-carboxyindole, 7-indolecarboxylic acid, acmc-209dvl, indole 7-carboxylic acid, indole-7-carboxlic acid, indole-7-carboxylic acid, indole-7-carboxylicacid, pubchem2475, zlchem 444
1H-indole-7-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 216-801-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents