missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Indole-7-carboxaldehyde, 98%, ACROS Organics™

Chemical Identifiers

CAS 1074-88-0
Assay Percent Range 98%
Molecular Formula C9H7NO
Formula Weight 145.16
MDL Number MFCD01318152
Chemical Name or Material Indole-7-carboxaldehyde
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC366620010 SDS Thermo Fisher Scientific
AC366620010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

1074-88-0
C9H7NO
MFCD01318152
≥97.5%
98%
145.16
Indole-7-carboxaldehyde

Specifications

145.16
1g
86° to 90°C
Glass bottle
C9H7NO
1074-88-0
≥97.5%
Crystalline Powder
Yellow
Authentic
98%
MFCD01318152
Indole-7-carboxaldehyde

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.