missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Indole-6-carboxylic Acid, TCI America™

$64.17 - $197.09

Chemical Identifiers

CAS 1670-82-2
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD00210441
InChI Key GHTDODSYDCPOCW-UHFFFAOYSA-N
Synonym indole-6-carboxylic acid, 6-indolecarboxylic acid, 6-carboxyindole, 6-indole carboxylic acid, pubchem1695, 6-carboxy-1h-indole, indole-6-carboxylicacid, indol-6-carboxylic acid, indole 6-carboxylic acid, maybridge4_003509
PubChem CID 595230
IUPAC Name 1H-indole-6-carboxylic acid
SMILES C1=CC(=CC2=C1C=CN2)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I06631G View Documents TCI America
I06631G
1g Each for $64.17
 
I06635G View Documents TCI America
I06635G
5g Each for $197.09
 
Specifications

Chemical Identifiers

1670-82-2
161.16
GHTDODSYDCPOCW-UHFFFAOYSA-N
595230
C1=CC(=CC2=C1C=CN2)C(=O)O
C9H7NO2
MFCD00210441
indole-6-carboxylic acid, 6-indolecarboxylic acid, 6-carboxyindole, 6-indole carboxylic acid, pubchem1695, 6-carboxy-1h-indole, indole-6-carboxylicacid, indol-6-carboxylic acid, indole 6-carboxylic acid, maybridge4_003509
1H-indole-6-carboxylic acid

Specifications

Undesignated
1g
C9H7NO2
indole-6-carboxylic acid, 6-indolecarboxylic acid, 6-carboxyindole, 6-indole carboxylic acid, pubchem1695, 6-carboxy-1h-indole, indole-6-carboxylicacid, indol-6-carboxylic acid, indole 6-carboxylic acid, maybridge4_003509
C1=CC(=CC2=C1C=CN2)C(=O)O
161.16
161.16
≥98.0% (HPLC,T)
257°C
1670-82-2
MFCD00210441
GHTDODSYDCPOCW-UHFFFAOYSA-N
1H-indole-6-carboxylic acid
595230
Crystal-Powder at 20°C
Indole-6-carboxylic Acid
Safety and Handling

Safety and Handling

TSCA : No