Please login to your online account to display your discounted pricing

Alfa Aesar™ Indole-6-carboxylic acid, 98%

$24.00 - $207.80

Chemical Identifiers

Chemical Name or Material Indole-6-carboxylic acid
CAS 1670-82-2
Molecular Formula C9H7NO2
Formula Weight 161.16
MDL Number MFCD00210441
Beilstein 123991
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1601906

  SDS
alfa aesar™
L1601906
5g Each for $207.80

AAL1601903

  SDS
alfa aesar™
L1601903
1g Each for $65.88

AAL16019MD

  SDS
alfa aesar™
L16019MD
250mg Each for $24.00
Description & Specifications

Specifications

Formula Weight 161.16
Chemical Name or Material Indole-6-carboxylic acid
Assay 98%
Molecular Formula C9H7NO2
MDL Number MFCD00210441
CAS 1670-82-2
Beilstein 123991
Health Hazard 1 Warning
Melting Point ≈258°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.