missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Indole-6-carboxaldehyde, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC435512500

 View more versions of this product

Catalog No. AC435512500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Indole-6-carboxaldehyde
Glass bottle
129°C to 131°C
1196-70-9
C9H7NO
1h-indole-6-carboxaldehyde, 6-formyl indole, 6-formyl-1h-indole, 6-formyl-indole, 6-formylindole, 6-indolecarboxaldehyde, indole-6-carbaldehyde, indole-6-carboxaldehyde, indole-6-carboxaldehyde 6-formylindole, indole-6-carboxalehyde
C1=CC(=CC2=C1C=CN2)C=O
145.16
145.16
97%
Authentic
Brown to Yellow
250mg
96% min. (HPLC)
MFCD02179596
VSPBWOAEHQDXRD-UHFFFAOYSA-N
1H-indole-6-carbaldehyde
2773435
Crystalline Powder

Chemical Identifiers

1196-70-9
145.16
VSPBWOAEHQDXRD-UHFFFAOYSA-N
2773435
C1=CC(=CC2=C1C=CN2)C=O
C9H7NO
MFCD02179596
1h-indole-6-carboxaldehyde, 6-formyl indole, 6-formyl-1h-indole, 6-formyl-indole, 6-formylindole, 6-indolecarboxaldehyde, indole-6-carbaldehyde, indole-6-carboxaldehyde, indole-6-carboxaldehyde 6-formylindole, indole-6-carboxalehyde
1H-indole-6-carbaldehyde
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

SDS Documents