missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Indole-6-boronic acid, 98%

Manufacturer:  Alfa Aesar™ H5331303

 View more versions of this product

Catalog No. AAH5331303



Specifications

Specifications

Indole-6-boronic acid
1g
C8H8BNO2
indole-6-boronic acid, 6-indolylboronic acid, 1h-indol-6-yl boronic acid, 6-indoleboronic acid, indole-6-boronicacid, 1h-indol-6-yl-6-boronic acid, 6-borono-1h-indole, 6-indole boronic acid, 6-boronoindole, 1h-indole-6-boronic acid
B(C1=CC2=C(C=C1)C=CN2)(O)O
160.967
160.97
177°C to 181°C
147621-18-9
MFCD03095176
ZVMHOIWRCCZGPZ-UHFFFAOYSA-N
1H-indol-6-ylboronic acid
2763205
98%

Chemical Identifiers

147621-18-9
160.967
ZVMHOIWRCCZGPZ-UHFFFAOYSA-N
2763205
B(C1=CC2=C(C=C1)C=CN2)(O)O
C8H8BNO2
MFCD03095176
indole-6-boronic acid, 6-indolylboronic acid, 1h-indol-6-yl boronic acid, 6-indoleboronic acid, indole-6-boronicacid, 1h-indol-6-yl-6-boronic acid, 6-borono-1h-indole, 6-indole boronic acid, 6-boronoindole, 1h-indole-6-boronic acid
1H-indol-6-ylboronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : −30° to −10°C

SDS Documents