missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Indole-5-carboxaldehyde 98.0+%, TCI America™
$202.40 - $807.94
Chemical Identifiers
CAS | 1196-69-6 |
---|---|
Molecular Formula | C9H7NO |
Molecular Weight (g/mol) | 145.161 |
MDL Number | MFCD02093664 |
InChI Key | ADZUEEUKBYCSEY-UHFFFAOYSA-N |
Synonym | indole-5-carboxaldehyde, 5-formylindole, indole-5-carbaldehyde, 1h-indole-5-carboxaldehyde, 5-indolealdehyde, indole-5-aldehyde, 5-formyl-1h-indole, 5-indolecarboxaldehyde, indole-5-carboxaldehyde 5-formylindole, 5-formyl-indole |
PubChem CID | 589040 |
IUPAC Name | 1H-indole-5-carbaldehyde |
SMILES | C1=CC2=C(C=CN2)C=C1C=O |
Chemical Identifiers
1196-69-6 | |
145.161 | |
ADZUEEUKBYCSEY-UHFFFAOYSA-N | |
589040 | |
C1=CC2=C(C=CN2)C=C1C=O |
C9H7NO | |
MFCD02093664 | |
indole-5-carboxaldehyde, 5-formylindole, indole-5-carbaldehyde, 1h-indole-5-carboxaldehyde, 5-indolealdehyde, indole-5-aldehyde, 5-formyl-1h-indole, 5-indolecarboxaldehyde, indole-5-carboxaldehyde 5-formylindole, 5-formyl-indole | |
1H-indole-5-carbaldehyde |
Specifications
1196-69-6 | |
C9H7NO | |
5 g | |
ADZUEEUKBYCSEY-UHFFFAOYSA-N | |
1H-indole-5-carbaldehyde | |
589040 | |
≥98.0% (GC) | |
Indole-5-carboxaldehyde |
100°C | |
MFCD02093664 | |
indole-5-carboxaldehyde, 5-formylindole, indole-5-carbaldehyde, 1h-indole-5-carboxaldehyde, 5-indolealdehyde, indole-5-aldehyde, 5-formyl-1h-indole, 5-indolecarboxaldehyde, indole-5-carboxaldehyde 5-formylindole, 5-formyl-indole | |
C1=CC2=C(C=CN2)C=C1C=O | |
145.161 | |
145.16 | |
Crystalline Powder |
Safety and Handling
TSCA : No