Please login to your online account to display your discounted pricing

Alfa Aesar™ Indole-5-carboxaldehyde, 98%

$58.31 - $223.00

Chemical Identifiers

Chemical Name or Material Indole-5-carboxaldehyde
CAS 1196-69-6
Molecular Formula C9H7NO
Formula Weight 145.16
MDL Number MFCD02093664
Synonym 5-Formylindole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2551703

  SDS
alfa aesar™
B2551703
1g Each for $58.31

AAB2551706

  SDS
alfa aesar™
B2551706
5g Each for $223.00
Description & Specifications

Specifications

Formula Weight 145.16
Synonym 5-Formylindole
Chemical Name or Material Indole-5-carboxaldehyde
Melting Point 96° to 100°C
MDL Number MFCD02093664
Quantity 1g
CAS 1196-69-6
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 98%
Molecular Formula C9H7NO
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Applications
Indole-5-carboxaldehyde is used as a reactant in preparation of curcumin derivatives as anti-proliferative & anti-inflammatory agents; and in synthesis of para-para stilbenophanes by McMurry coupling. It is also used as a reactant in stereoselective synthesis of dibenzylideneacetone derivatives as β-amyloid imaging probes and in structure-based drug design of aurora kinase A inhibitors.

Solubility
Insoluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.