Please login to your online account to display your discounted pricing

Alfa Aesar™ Indole-5-carbonitrile, 98+%

$80.35 - $1,125.22

Chemical Identifiers

Chemical Name or Material Indole-5-carbonitrile
CAS 15861-24-2
Molecular Formula C9H6N2
Formula Weight 142.16
MDL Number MFCD00005669
Beilstein 116738
Synonym 5-Cyanoindole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2181003

  SDS
alfa aesar™
B2181003
1g Each for $80.35

AAB2181006

  SDS
alfa aesar™
B2181006
5g Each for $270.92

AAB2181014

  SDS
alfa aesar™
B2181014
25g Each for $1,125.22
Description & Specifications

Specifications

Formula Weight 142.16
Chemical Name or Material Indole-5-carbonitrile
Assay ≥98%
Molecular Formula C9H6N2
MDL Number MFCD00005669
CAS 15861-24-2
Beilstein 116738
Synonym 5-Cyanoindole
Health Hazard 1 Warning
Melting Point 104° to 108°C
Quantity 1g
Odor Odorless
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number NL5993120

Applications
Reactant used for parallel synthesis of dihydroisoquinolines via silver and L-proline co-catalyzed three-component coupling reaction, preparation of novel PPARα/γ dual agonists for potential treatment of metabolic syndrome and IDDM, chemo selective and regioselective preparation of benzoyl indoles, 4,5-dihydrocyclopenta[c]quinolines by palladium-catalyzed ring-expansion reaction alkynes, using O2 as the oxidant and vinylindoles by hydroarylation of alkynes using indium bromide catalyst.

Solubility
Soluble in chloroform, hexane and methanol. Insoluble in water.

Notes
Light Sensitive, store in dark. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.