Please login to your online account to display your discounted pricing

Alfa Aesar™ Indole-5-boronic acid, 95%

$113.91 - $113.91

Chemical Identifiers

Chemical Name or Material Indole-5-boronic acid
CAS 144104-59-6
Molecular Formula C8H8BNO2
Formula Weight 160.97
MDL Number MFCD01319013
Synonym 5-Boronoindole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2700803

  SDS
alfa aesar™
H2700803
1g Each for $113.91
Description & Specifications

Specifications

Formula Weight 160.97
Synonym 5-Boronoindole
Assay 95%
MDL Number MFCD01319013
Melting Point 170° to 175°C
Quantity 1g
Molecular Formula C8H8BNO2
CAS 144104-59-6
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Recommended Storage 2° to 8°C
Chemical Name or Material Indole-5-boronic acid
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C