missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Indole-4-carboxylic Acid, TCI America™

$204.72 - $678.34

Chemical Identifiers

CAS 2124-55-2
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD00009739
InChI Key ROGHUJUFCRFUSO-UHFFFAOYSA-N
Synonym indole-4-carboxylic acid, indole-4-carboxylicacid, 4-carboxyindole, 1h-4-indolecarboxylic acid, 1h-indole-4-carboxylic?acid, 4-indolecarboxylic acid, pubchem1693, acmc-209fio, 4-indole-carboxylic acid, indol-4-yl carboxylic acid
PubChem CID 595229
IUPAC Name 1H-indole-4-carboxylic acid
SMILES C1=CC(=C2C=CNC2=C1)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I07221G View Documents TCI America
I07221G
1g Each for $204.72
 
I07225G View Documents TCI America
I07225G
5g Each for $678.34
 
Specifications

Chemical Identifiers

2124-55-2
161.16
ROGHUJUFCRFUSO-UHFFFAOYSA-N
595229
C1=CC(=C2C=CNC2=C1)C(=O)O
C9H7NO2
MFCD00009739
indole-4-carboxylic acid, indole-4-carboxylicacid, 4-carboxyindole, 1h-4-indolecarboxylic acid, 1h-indole-4-carboxylic?acid, 4-indolecarboxylic acid, pubchem1693, acmc-209fio, 4-indole-carboxylic acid, indol-4-yl carboxylic acid
1H-indole-4-carboxylic acid

Specifications

Undesignated
1g
C9H7NO2
indole-4-carboxylic acid, indole-4-carboxylicacid, 4-carboxyindole, 1h-4-indolecarboxylic acid, 1h-indole-4-carboxylic?acid, 4-indolecarboxylic acid, pubchem1693, acmc-209fio, 4-indole-carboxylic acid, indol-4-yl carboxylic acid
C1=CC(=C2C=CNC2=C1)C(=O)O
161.16
161.16
≥98.0% (GC,T)
212°C
2124-55-2
MFCD00009739
ROGHUJUFCRFUSO-UHFFFAOYSA-N
1H-indole-4-carboxylic acid
595229
Crystal-Powder at 20°C
Indole-4-carboxylic Acid
Safety and Handling

Safety and Handling

TSCA : No