missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Indole-4-carboxylic acid, 98%, ACROS Organics™

Chemical Identifiers

CAS 2124-55-2
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD00009739
InChI Key ROGHUJUFCRFUSO-UHFFFAOYSA-N
Synonym 1h-4-indolecarboxylic acid, 1h-indole-4-carboxylic?acid, 4-carboxyindole, 4-indole-carboxylic acid, 4-indolecarboxylic acid, acmc-209fio, indol-4-yl carboxylic acid, indole-4-carboxylic acid, indole-4-carboxylicacid, pubchem1693
PubChem CID 595229
IUPAC Name 1H-indole-4-carboxylic acid
SMILES C1=CC(=C2C=CNC2=C1)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC398060010
SDS View Documents
Thermo Fisher Scientific
AC398060010
1g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC398060050
SDS View Documents
Thermo Fisher Scientific
AC398060050
5g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

2124-55-2
161.16
ROGHUJUFCRFUSO-UHFFFAOYSA-N
595229
C1=CC(=C2C=CNC2=C1)C(=O)O
C9H7NO2
MFCD00009739
1h-4-indolecarboxylic acid, 1h-indole-4-carboxylic?acid, 4-carboxyindole, 4-indole-carboxylic acid, 4-indolecarboxylic acid, acmc-209fio, indol-4-yl carboxylic acid, indole-4-carboxylic acid, indole-4-carboxylicacid, pubchem1693
1H-indole-4-carboxylic acid

Specifications

Authentic
Beige
1g
97.5% min. (HPLC)
MFCD00009739
ROGHUJUFCRFUSO-UHFFFAOYSA-N
1H-indole-4-carboxylic acid
595229
Powder
Indole-4-carboxylic acid
Glass bottle
207°C to 209°C
2124-55-2
C9H7NO2
1h-4-indolecarboxylic acid, 1h-indole-4-carboxylic?acid, 4-carboxyindole, 4-indole-carboxylic acid, 4-indolecarboxylic acid, acmc-209fio, indol-4-yl carboxylic acid, indole-4-carboxylic acid, indole-4-carboxylicacid, pubchem1693
C1=CC(=C2C=CNC2=C1)C(=O)O
161.16
161.16
98%
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning