missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Indole-4-carboxylic acid, 97%

Manufacturer:  Alfa Aesar™ H55092MD

 View more versions of this product

Catalog No. AAH55092MD



Specifications

Specifications

Indole-4-carboxylic acid
250mg
C9H7NO2
1h-4-indolecarboxylic acid
C1=CC(=C2C=CNC2=C1)C(=O)O
161.16
161.16
213°C to 214°C
2124-55-2
MFCD00009739
ROGHUJUFCRFUSO-UHFFFAOYSA-N
1H-indole-4-carboxylic acid
595229
97%

Chemical Identifiers

2124-55-2
161.16
ROGHUJUFCRFUSO-UHFFFAOYSA-N
595229
C1=CC(=C2C=CNC2=C1)C(=O)O
C9H7NO2
MFCD00009739
1h-4-indolecarboxylic acid
1H-indole-4-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents