Please login to your online account to display your discounted pricing

Alfa Aesar™ Indole-4-boronic acid, 95%

$119.92 - $119.92

Chemical Identifiers

Chemical Name or Material Indole-4-boronic acid
CAS 220465-43-0
Molecular Formula C8H8BNO2
Formula Weight 160.97
MDL Number MFCD03095175
Synonym 4-Boronoindole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH53037MD

 
alfa aesar™
H53037MD
250mg Each for $119.92
Description & Specifications

Specifications

Formula Weight 160.97
Chemical Name or Material Indole-4-boronic acid
Assay 95%
Molecular Formula C8H8BNO2
MDL Number MFCD03095175
CAS 220465-43-0
Synonym 4-Boronoindole
Health Hazard 1 Warning
Quantity 250mg
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage −30° to −10°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

−30° to −10°C